missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-4-piperidinecarboxylic Acid 98.0+%, TCI America™
$74.56 - $240.46
Chemical Identifiers
| CAS | 25503-90-6 |
|---|---|
| Molecular Formula | C8H12NO3 |
| Molecular Weight (g/mol) | 170.19 |
| MDL Number | MFCD00023706 |
| InChI Key | WFCLWJHOKCQYOQ-UHFFFAOYSA-M |
| Synonym | 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl |
| PubChem CID | 117255 |
| IUPAC Name | 1-acetylpiperidine-4-carboxylate |
| SMILES | CC(=O)N1CCC(CC1)C([O-])=O |
Chemical Identifiers
| 25503-90-6 | |
| 170.19 | |
| WFCLWJHOKCQYOQ-UHFFFAOYSA-M | |
| 117255 | |
| CC(=O)N1CCC(CC1)C([O-])=O |
| C8H12NO3 | |
| MFCD00023706 | |
| 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl | |
| 1-acetylpiperidine-4-carboxylate |
Specifications
| 25503-90-6 | |
| White-Yellow | |
| MFCD00023706 | |
| 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl | |
| CC(=O)N1CCC(CC1)C([O-])=O | |
| 170.19 | |
| 171.20 | |
| Crystalline Powder |
| 184°C | |
| C8H12NO3 | |
| 5 g | |
| WFCLWJHOKCQYOQ-UHFFFAOYSA-M | |
| 1-acetylpiperidine-4-carboxylate | |
| 117255 | |
| ≥98.0% (GC,T) | |
| 1-Acetyl-4-piperidinecarboxylic Acid |
Safety and Handling
TSCA : No