accessibility menu, dialog, popup

UserName

H3CONOO

CAS RN 25503-90-6

IUPAC Name: 1-acetylpiperidine-4-carboxylate
Synonyms: 1-acetylpiperidine-4-carboxylate
Molecular Weight (g/mol): 170.19
Molecular Formula: C8H12NO3
InChi Key: WFCLWJHOKCQYOQ-UHFFFAOYSA-M
SMILES: CC(=O)N1CCC(CC1)C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)