missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-4-piperidinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America A226125G
Specifications
| 1-Acetyl-4-piperidinecarboxylic Acid | |
| 184°C | |
| C8H12NO3 | |
| 25 g | |
| WFCLWJHOKCQYOQ-UHFFFAOYSA-M | |
| 1-acetylpiperidine-4-carboxylate | |
| 117255 | |
| ≥98.0% (GC,T) |
| 25503-90-6 | |
| White-Yellow | |
| MFCD00023706 | |
| 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl | |
| CC(=O)N1CCC(CC1)C([O-])=O | |
| 170.19 | |
| 171.20 | |
| Crystalline Powder |
Chemical Identifiers
| 25503-90-6 | |
| 170.19 | |
| WFCLWJHOKCQYOQ-UHFFFAOYSA-M | |
| 117255 | |
| CC(=O)N1CCC(CC1)C([O-])=O |
| C8H12NO3 | |
| MFCD00023706 | |
| 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl | |
| 1-acetylpiperidine-4-carboxylate |
Safety and Handling
TSCA : No