missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-2-phenylhydrazine 98.0+%, TCI America™
Supplier: TCI America A010725G
Specifications
| 1-Acetyl-2-phenylhydrazine | |
| 130°C | |
| MFCD00008672 | |
| 2811 | |
| UICBCXONCUFSOI-UHFFFAOYSA-N | |
| N'-phenylacetohydrazide | |
| 8247 | |
| ≥98.0% (GC) |
| 114-83-0 | |
| C8H10N2O | |
| 25 g | |
| 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine | |
| CC(=O)NNC1=CC=CC=C1 | |
| 150.181 | |
| 150.18 | |
| Crystalline Powder |
Chemical Identifiers
| 114-83-0 | |
| 150.181 | |
| UICBCXONCUFSOI-UHFFFAOYSA-N | |
| 8247 | |
| CC(=O)NNC1=CC=CC=C1 |
| C8H10N2O | |
| MFCD00008672 | |
| 1-acetyl-2-phenylhydrazine, hydracetin, acetylphenylhydrazine, pyrodin, pyrodine, acetic acid, 2-phenylhydrazide, n-acetyl-n'-phenylhydrazine, n'-phenylacethydrazide, acetic acid phenylhydrazone, n 1-acetylphenylhydrazine | |
| N'-phenylacetohydrazide |
Safety and Handling
RTECSNumber : AJ2900000
TSCA : Yes