missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-1,2,3,4-tetrahydroquinoline 98.0+%, TCI America™
$130.48 - $326.21
Chemical Identifiers
| CAS | 4169-19-1 |
|---|---|
| Molecular Formula | C11H13NO |
| Molecular Weight (g/mol) | 175.23 |
| MDL Number | MFCD01195305 |
| InChI Key | RRWLNRQGJSQRAF-UHFFFAOYSA-N |
| Synonym | 1,2,3,4-Tetrahydro-1-acetylquinoline |
| PubChem CID | 347850 |
| IUPAC Name | 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one |
| SMILES | CC(=O)N1CCCC2=CC=CC=C12 |
Chemical Identifiers
| 4169-19-1 | |
| 175.23 | |
| RRWLNRQGJSQRAF-UHFFFAOYSA-N | |
| 347850 | |
| CC(=O)N1CCCC2=CC=CC=C12 |
| C11H13NO | |
| MFCD01195305 | |
| 1,2,3,4-Tetrahydro-1-acetylquinoline | |
| 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one |
Specifications
| 4169-19-1 | |
| 295°C | |
| MFCD01195305 | |
| 1,2,3,4-Tetrahydro-1-acetylquinoline | |
| CC(=O)N1CCCC2=CC=CC=C12 | |
| 175.23 | |
| 175.23 | |
| Liquid |
| Yellow | |
| C11H13NO | |
| 5 g | |
| RRWLNRQGJSQRAF-UHFFFAOYSA-N | |
| 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one | |
| 347850 | |
| ≥98.0% (GC) | |
| 1-Acetyl-1,2,3,4-tetrahydroquinoline |
Safety and Handling
TSCA : No