accessibility menu, dialog, popup

UserName

CAS RN 4169-19-1

H3CON

CAS RN 4169-19-1

IUPAC Name: 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one
Synonyms: 1-(3,4-dihydro-2H-quinolin-1-yl)ethanone
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: RRWLNRQGJSQRAF-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results