missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-1,2,3,4-tetrahydroquinoline 98.0+%, TCI America™
Supplier: TCI America A164125G
Specifications
| 1-Acetyl-1,2,3,4-tetrahydroquinoline | |
| Yellow | |
| C11H13NO | |
| 25 g | |
| RRWLNRQGJSQRAF-UHFFFAOYSA-N | |
| 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one | |
| 347850 | |
| ≥98.0% (GC) |
| 4169-19-1 | |
| 295°C | |
| MFCD01195305 | |
| 1,2,3,4-Tetrahydro-1-acetylquinoline | |
| CC(=O)N1CCCC2=CC=CC=C12 | |
| 175.23 | |
| 175.23 | |
| Liquid |
Chemical Identifiers
| 4169-19-1 | |
| 175.23 | |
| RRWLNRQGJSQRAF-UHFFFAOYSA-N | |
| 347850 | |
| CC(=O)N1CCCC2=CC=CC=C12 |
| C11H13NO | |
| MFCD01195305 | |
| 1,2,3,4-Tetrahydro-1-acetylquinoline | |
| 1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one |
Safety and Handling
TSCA : No