missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-2-methoxy-4-[(E)-1-propenyl]benzene 98.0+%, TCI America™

Catalog No. A118425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A118425G 25 g
A1184500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A118425G Supplier TCI America Supplier No. A118425G
Only null left

Chemical Identifiers

CAS 93-29-8
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00026984
InChI Key IUSBVFZKQJGVEP-SNAWJCMRSA-N
Synonym Acetylisoeugenol
PubChem CID 876160
ChEBI CHEBI:86583
IUPAC Name 2-methoxy-4-[(1E)-prop-1-en-1-yl]phenyl acetate
SMILES COC1=CC(\C=C\C)=CC=C1OC(C)=O

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 283°C
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetoxy-2-methoxy-4-(1-propenyl)benzene
RTECSNumber SL7940000
TSCA Yes