missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic Acid 95.0+%, TCI America™
Supplier: TCI America F08665G
Specifications
| 1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic Acid | |
| 187°C | |
| C21H21NO4 | |
| 5 g | |
| IECZEINPZOFWNU-UHFFFAOYSA-N | |
| 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentane-1-carboxylic acid | |
| 978345 | |
| ≥95.0% (HPLC,T) |
| 117322-30-2 | |
| White-Yellow | |
| MFCD01074696 | |
| 1-(Fmoc-amino)cyclopentanecarboxylic Acid, Fmoc-cycloleucine | |
| C1CCC(C1)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
| 351.402 | |
| 351.40 | |
| Crystalline Powder |
Chemical Identifiers
| 117322-30-2 | |
| 351.402 | |
| IECZEINPZOFWNU-UHFFFAOYSA-N | |
| 978345 | |
| C1CCC(C1)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
| C21H21NO4 | |
| MFCD01074696 | |
| 1-(Fmoc-amino)cyclopentanecarboxylic Acid, Fmoc-cycloleucine | |
| 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentane-1-carboxylic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator