missing translation for 'onlineSavingsMsg'
Learn More

1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic Acid 95.0+%, TCI America™

Catalog No. p-7129948
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08661G 1 g
F08665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08661G Supplier TCI America Supplier No. F08661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117322-30-2
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD01074696
InChI Key IECZEINPZOFWNU-UHFFFAOYSA-N
Synonym 1-(Fmoc-amino)cyclopentanecarboxylic Acid, Fmoc-cycloleucine
PubChem CID 978345
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 1 g
Formula Weight 351.40
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]cyclopentanecarboxylic Acid
TSCA No
Recommended Storage Refrigerator