missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,9-Nonanediol 98.0+%, TCI America™
$89.23 - $617.05
Chemical Identifiers
| CAS | 3937-56-2 |
|---|---|
| Molecular Formula | C9H20O2 |
| Molecular Weight (g/mol) | 160.257 |
| MDL Number | MFCD00002991 |
| InChI Key | ALVZNPYWJMLXKV-UHFFFAOYSA-N |
| Synonym | 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x |
| PubChem CID | 19835 |
| IUPAC Name | nonane-1,9-diol |
| SMILES | C(CCCCO)CCCCO |
Chemical Identifiers
| C9H20O2 | |
| MFCD00002991 | |
| 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x | |
| nonane-1,9-diol |
Specifications
| 3937-56-2 | |
| White | |
| C9H20O2 | |
| 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x | |
| C(CCCCO)CCCCO | |
| 160.257 | |
| 160.26 | |
| Crystalline Powder |
| 46°C | |
| 171°C | |
| MFCD00002991 | |
| ALVZNPYWJMLXKV-UHFFFAOYSA-N | |
| nonane-1,9-diol | |
| 19835 | |
| ≥98.0% (GC) | |
| 1,9-Nonanediol |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : Yes