missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanediol 98.0+%, TCI America™

Catalog No. N028725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N028725G 25 g
N0287100G 100 g
N0287500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N028725G Supplier TCI America Supplier No. N028725G

Chemical Identifiers

CAS 3937-56-2
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD00002991
InChI Key ALVZNPYWJMLXKV-UHFFFAOYSA-N
Synonym 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
PubChem CID 19835
IUPAC Name nonane-1,9-diol
SMILES C(CCCCO)CCCCO

Specifications

Melting Point 46°C
Color White
Boiling Point 171°C
Quantity 25 g
Formula Weight 160.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,9-Nonanediol
EINECSNumber (2)-0240
TSCA Yes