accessibility menu, dialog, popup

UserName

HOOH

CAS RN 3937-56-2

IUPAC Name: nonane-1,9-diol
Synonyms: 1,9-nonanediol
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: ALVZNPYWJMLXKV-UHFFFAOYSA-N
SMILES: OCCCCCCCCCO
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)