missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Naphthosultone 98.0+%, TCI America™
Supplier: TCI America N076625G
Specifications
| 1,8-Naphthosultone | |
| 158°C | |
| C10H6O3S | |
| 8-Hydroxynaphthalene-1-sulfonic Acid Sultone, 1,8-Naphthalenesultone, 1-Naphthol-8-sulfonic Acid Sultone | |
| C1=CC2=C3C(=C1)OS(=O)(=O)C3=CC=C2 | |
| 65744 | |
| ≥98.0% (HPLC) |
| 83-31-8 | |
| Yellow | |
| 25 g | |
| IEIADDVJUYQKAZ-UHFFFAOYSA-N | |
| 206.215 | |
| 206.22 | |
| Crystalline Powder |
Chemical Identifiers
| 83-31-8 | |
| 206.215 | |
| 8-Hydroxynaphthalene-1-sulfonic Acid Sultone, 1,8-Naphthalenesultone, 1-Naphthol-8-sulfonic Acid Sultone | |
| C1=CC2=C3C(=C1)OS(=O)(=O)C3=CC=C2 |
| C10H6O3S | |
| IEIADDVJUYQKAZ-UHFFFAOYSA-N | |
| 65744 |
Safety and Handling
EINECSNumber : (5)-0534
TSCA : Yes