accessibility menu, dialog, popup

UserName

CAS RN 83-31-8

IUPAC Name: 2-oxa-3λ⁶-thiatricyclo[6.3.1.0⁴,¹²]dodeca-1(12),4,6,8,10-pentaene-3,3-dione
Synonyms: 2-oxa-3λ⁶-thiatricyclo[6.3.1.0⁴,¹²]dodeca-1(12),4,6,8,10-pentaene-3,3-dione
Molecular Weight (g/mol): 206.22
Molecular Formula: C10H6O3S
InChi Key: IEIADDVJUYQKAZ-UHFFFAOYSA-N
SMILES: O=S1(=O)OC2=C3C1=CC=CC3=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results