missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthosultone 98.0+%, TCI America™

Catalog No. N07665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07665G 5 g
N076625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07665G Supplier TCI America Supplier No. N07665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83-31-8
Molecular Formula C10H6O3S
Molecular Weight (g/mol) 206.215
InChI Key IEIADDVJUYQKAZ-UHFFFAOYSA-N
Synonym 8-Hydroxynaphthalene-1-sulfonic Acid Sultone, 1,8-Naphthalenesultone, 1-Naphthol-8-sulfonic Acid Sultone
PubChem CID 65744
SMILES C1=CC2=C3C(=C1)OS(=O)(=O)C3=CC=C2

Specifications

Melting Point 158°C
Color Yellow
Quantity 5 g
Formula Weight 206.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Naphthosultone
EINECSNumber (5)-0534
TSCA Yes