missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Naphthalic Anhydride 98.0+%, TCI America™
$55.84 - $100.11
Chemical Identifiers
| CAS | 81-84-5 |
|---|---|
| Molecular Formula | C12H6O3 |
| Molecular Weight (g/mol) | 198.18 |
| MDL Number | MFCD00006925 |
| InChI Key | GRSMWKLPSNHDHA-UHFFFAOYSA-N |
| Synonym | 1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione |
| PubChem CID | 6693 |
| ChEBI | CHEBI:82246 |
| IUPAC Name | 3-oxatricyclo[7.3.1.0âµ,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione |
| SMILES | O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2 |
Chemical Identifiers
| 81-84-5 | |
| 198.18 | |
| GRSMWKLPSNHDHA-UHFFFAOYSA-N | |
| 6693 | |
| 3-oxatricyclo[7.3.1.0âµ,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione |
| C12H6O3 | |
| MFCD00006925 | |
| 1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione | |
| CHEBI:82246 | |
| O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2 |
Specifications
| 81-84-5 | |
| Yellow | |
| MFCD00006925 | |
| 1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione | |
| O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2 | |
| 198.18 | |
| CHEBI:82246 | |
| ≥98.0% (T,HPLC) | |
| 1,8-Naphthalic Anhydride |
| 274°C | |
| C12H6O3 | |
| 25 g | |
| GRSMWKLPSNHDHA-UHFFFAOYSA-N | |
| 3-oxatricyclo[7.3.1.0âµ,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione | |
| 6693 | |
| 198.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0404
RTECSNumber : QK5350000
TSCA : Yes