missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalic Anhydride 98.0+%, TCI America™

Supplier:  TCI America N0022250G

 View more versions of this product

Catalog No. N0022250G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,8-Naphthalic Anhydride
274°C
C12H6O3
250 g
GRSMWKLPSNHDHA-UHFFFAOYSA-N
3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
6693
198.18
Crystalline Powder
81-84-5
Yellow
MFCD00006925
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2
198.18
CHEBI:82246
≥98.0% (T,HPLC)

Chemical Identifiers

81-84-5
198.18
GRSMWKLPSNHDHA-UHFFFAOYSA-N
6693
3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
C12H6O3
MFCD00006925
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
CHEBI:82246
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0404

RTECSNumber : QK5350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions