missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dibromooctane, 98%
CAS: 4549-32-0 | C8H16Br2 | 272.01 g/mol
Supplier: Thermo Scientific Chemicals 112880500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 8-Dibromooctane | |
| 97.5 | |
| 15.0°C to 16.0°C | |
| 1.4600g/mL | |
| >112°C | |
| 98% | |
| C8H16Br2 | |
| Br(CH2)8Br | |
| 01,16 | |
| octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r | |
| DKEGCUDAFWNSSO-UHFFFAOYSA-N | |
| 1,8-dibromooctane | |
| 78310 | |
| 98% |
| 4549-32-0 | |
| 100.0 | |
| Yellow | |
| 270.0°C to 272.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4981 to 1.5001 | |
| 50 mL | |
| 1.46 | |
| Solubility in water: immiscible | |
| C(CCCCBr)CCCBr | |
| 272.01 | |
| 272.01 | |
| Liquid |
Chemical Identifiers
| 4549-32-0 | |
| 272.01 | |
| octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r | |
| 1,8-dibromooctane |
| C8H16Br2 | |
| DKEGCUDAFWNSSO-UHFFFAOYSA-N | |
| 78310 | |
| C(CCCCBr)CCCBr |
Safety and Handling
EINECSNumber : 224-912-5
RUO – Research Use Only