missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, 98%

Catalog No. p-4258928
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112880100 10 mL Glass bottle
AC112880500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112880100 Supplier Thermo Scientific Chemicals Supplier No. 112880100
Only null left

CAS: 4549-32-0 | C8H16Br2 | 272.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
Molecular Weight (g/mol) 272.01
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane
SMILES C(CCCCBr)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 15.0°C to 16.0°C
Color Yellow
Density 1.4600g/mL
Boiling Point 270.0°C to 272.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4981 to 1.5001
Linear Formula Br(CH2)8Br
Quantity 10 mL
Beilstein 01,16
Specific Gravity 1.46
Solubility Information Solubility in water: immiscible
Formula Weight 272.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 8-Dibromooctane
Show More Show Less
EINECSNumber 224-912-5

RUO – Research Use Only