missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane

Catalog No. AAH26643MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26643MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26643MD Supplier Thermo Scientific Chemicals Supplier No. H26643MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 214078-93-0 | C24H36N4 | 380.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214078-93-0
Molecular Formula C24H36N4
Molecular Weight (g/mol) 380.58
MDL Number MFCD09263312
InChI Key QWGFFCRTIFBAFJ-UHFFFAOYSA-N
Synonym 1,8-dibenzyl-1,4,8,11-tetraazacyclotetradecane, acmc-1cidi, n 1 ,n 8-dibenzylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,8-bis phenylmethyl
PubChem CID 11176466
IUPAC Name 1,8-dibenzyl-1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCN(CCCNCCN(C1)CC2=CC=CC=C2)CC3=CC=CC=C3

Specifications

Melting Point 67°C to 68°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 380.58
Chemical Name or Material 1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only