accessibility menu, dialog, popup

UserName

CAS RN 214078-93-0

IUPAC Name: 1,8-dibenzyl-1,4,8,11-tetraazacyclotetradecane
Synonyms: 1,8-dibenzyl-1,4,8,11-tetraazacyclotetradecane
Molecular Weight (g/mol): 380.58
Molecular Formula: C24H36N4
InChi Key: QWGFFCRTIFBAFJ-UHFFFAOYSA-N
SMILES: C(N1CCCNCCN(CC2=CC=CC=C2)CCCNCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results