missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Diaminonaphthalene 98.0+%, TCI America™
$205.05 - $1998.27
Chemical Identifiers
| CAS | 479-27-6 |
|---|---|
| Molecular Formula | C10H10N2 |
| Molecular Weight (g/mol) | 158.204 |
| MDL Number | MFCD00004033 |
| InChI Key | YFOOEYJGMMJJLS-UHFFFAOYSA-N |
| Synonym | 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t |
| PubChem CID | 68067 |
| IUPAC Name | naphthalene-1,8-diamine |
| SMILES | C1=CC2=C(C(=C1)N)C(=CC=C2)N |
Chemical Identifiers
| 479-27-6 | |
| 158.204 | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| 68067 | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N |
| C10H10N2 | |
| MFCD00004033 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| naphthalene-1,8-diamine |
Specifications
| 479-27-6 | |
| 205°C | |
| MFCD00004033 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N | |
| 158.204 | |
| 158.20 | |
| Crystalline Powder |
| 65°C | |
| C10H10N2 | |
| 25 g | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| naphthalene-1,8-diamine | |
| 68067 | |
| ≥98.0% (T) | |
| 1,8-Diaminonaphthalene |
Safety and Handling
EINECSNumber : (4)-0324
TSCA : Yes