missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Diaminonaphthalene 98.0+%, TCI America™
Supplier: TCI America D0102500G
Specifications
| 1,8-Diaminonaphthalene | |
| 65°C | |
| C10H10N2 | |
| 500 g | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| naphthalene-1,8-diamine | |
| 68067 | |
| ≥98.0% (T) |
| 479-27-6 | |
| 205°C | |
| MFCD00004033 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N | |
| 158.204 | |
| 158.20 | |
| Crystalline Powder |
Chemical Identifiers
| 479-27-6 | |
| 158.204 | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| 68067 | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N |
| C10H10N2 | |
| MFCD00004033 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| naphthalene-1,8-diamine |
Safety and Handling
EINECSNumber : (4)-0324
TSCA : Yes