missing translation for 'onlineSavingsMsg'
Learn More

1,8-Bis(diphenylphosphino)naphthalene 97.0+%, TCI America™

Catalog No. B45305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45305G 5 g
B45301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45305G Supplier TCI America Supplier No. B45305G
Only null left

Chemical Identifiers

CAS 153725-04-3
Molecular Formula C34H26P2
Molecular Weight (g/mol) 496.53
MDL Number MFCD04117326
InChI Key QOXZKPURCFVBRR-UHFFFAOYSA-N
PubChem CID 2756729
IUPAC Name [8-(diphenylphosphanyl)naphthalen-1-yl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC2=CC=CC(P(C3=CC=CC=C3)C3=CC=CC=C3)=C12

Specifications

Melting Point 210°C
Color Yellow
Quantity 5 g
Formula Weight 496.53
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(diphenylphosphino)naphthalene