accessibility menu, dialog, popup

UserName

PP

CAS RN 153725-04-3

IUPAC Name: [8-(diphenylphosphanyl)naphthalen-1-yl]diphenylphosphane
Synonyms: [8-(diphenylphosphanyl)naphthalen-1-yl]diphenylphosphane
Molecular Weight (g/mol): 496.53
Molecular Formula: C34H26P2
InChi Key: QOXZKPURCFVBRR-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC2=CC=CC(P(C3=CC=CC=C3)C3=CC=CC=C3)=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results