missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dihydroxynaphthalene 98.0+%, TCI America™

Catalog No. p-7126307
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059225G 25 g
D0592500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D059225G Supplier TCI America Supplier No. D059225G
Only null left

Chemical Identifiers

CAS 575-38-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00035720
InChI Key ZUVBIBLYOCVYJU-UHFFFAOYSA-N
Synonym 1,7-dihydroxynaphthalene, 1,7-naphthalenediol, napthalene-1,7-diol, unii-7d42f605cs, ccris 7895, 1,7 dihydroxynaphthalene, 2,8-dihydroxynaphthalene, acmc-1aqz2, unii-m7uz7bfo3a component
PubChem CID 68462
IUPAC Name naphthalene-1,7-diol
SMILES C1=CC2=C(C=C(C=C2)O)C(=C1)O

Specifications

Melting Point 175°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,7-Dihydroxynaphthalene
EINECSNumber (4)-0356
RTECSNumber QJ4746666
TSCA Yes