missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,7-Dihydroxynaphthalene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 175°C |
| Quantity | 500 g |
| Formula Weight | 160.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,7-Dihydroxynaphthalene |
Safety and Handling
| EINECSNumber | (4)-0356 |
| RTECSNumber | QJ4746666 |
| TSCA | Yes |