missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Hexanediol 97.0+%, TCI America™
Chemical Identifiers
| CAS | 629-11-8 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.176 |
| MDL Number | MFCD00002985 |
| InChI Key | XXMIOPMDWAUFGU-UHFFFAOYSA-N |
| Synonym | 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol |
| PubChem CID | 12374 |
| ChEBI | CHEBI:43078 |
| IUPAC Name | hexane-1,6-diol |
| SMILES | C(CCCO)CCO |
Chemical Identifiers
| C6H14O2 | |
| MFCD00002985 | |
| 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol | |
| CHEBI:43078 | |
| C(CCCO)CCO |
Specifications
| 629-11-8 | |
| White | |
| C6H14O2 | |
| 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol | |
| C(CCCO)CCO | |
| 118.176 | |
| CHEBI:43078 | |
| ≥97.0% (GC) | |
| 1,6-Hexanediol |
| 42°C | |
| 208°C | |
| MFCD00002985 | |
| XXMIOPMDWAUFGU-UHFFFAOYSA-N | |
| hexane-1,6-diol | |
| 12374 | |
| 118.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0240
RTECSNumber : MO2100000
TSCA : Yes