missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 1,6-Hexanediol
Description
1,6-Hexanediol is a widely used tool to probe liquid-liquid phase separation (LLPS) in cells. Disrupts hydrophobic interaction-induced phase separation assemblies both in vitro and in vivo. Dissolves LLPS condensates, such as P bodies, but not solid-like condensates, such as protein aggregates and cytoskeletal assemblies.Typically used as a 3-10% solution (w/v). Disrupts cellular structures in yeast and alters cellular morphology in HeLa cells.
Specifications
Specifications
| CAS | 629-11-8 |
| Quantity | 10 g |
| Product Line | Tocris |
| Molecular Formula | C6H14O2 |
| Purity | 98% |