missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Dibromohexane 97.0+%, TCI America™
Supplier: TCI America D0185500G
Specifications
| 1,6-Dibromohexane | |
| -2°C | |
| 246°C | |
| MFCD00000272 | |
| hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane | |
| C(CCCBr)CCBr | |
| 243.97 | |
| 243.97 | |
| Liquid |
| 629-03-8 | |
| Yellow | |
| C6H12Br2 | |
| 500 g | |
| SGRHVVLXEBNBDV-UHFFFAOYSA-N | |
| 1,6-dibromohexane | |
| 12368 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 629-03-8 | |
| 243.97 | |
| SGRHVVLXEBNBDV-UHFFFAOYSA-N | |
| 12368 | |
| C(CCCBr)CCBr |
| C6H12Br2 | |
| MFCD00000272 | |
| hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane | |
| 1,6-dibromohexane |
Safety and Handling
RTECSNumber : MO1515000
TSCA : Yes