missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane 97.0+%, TCI America™

Catalog No. p-7126075
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D018525G 25 g
D0185100G 100 g
D0185500G 500 g
3 options

Catalog No. D018525G

Supplier: TCI America D018525G

Only null left

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 246°C
Quantity 25 g
Formula Weight 243.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dibromohexane
RTECSNumber MO1515000
TSCA Yes