missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Naphthalenedisulfonic Acid Tetrahydrate 98.0+%, TCI America™
$63.38 - $376.54
Chemical Identifiers
| CAS | 211366-30-2 |
|---|---|
| Molecular Formula | C10H16O10S2 |
| Molecular Weight (g/mol) | 360.35 |
| MDL Number | MFCD00149265 |
| InChI Key | ZLBLYGIIADHDKG-UHFFFAOYSA-N |
| PubChem CID | 53249422 |
| IUPAC Name | naphthalene-1,5-disulfonic acid tetrahydrate |
| SMILES | O.O.O.O.OS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(O)(=O)=O |
Chemical Identifiers
| 211366-30-2 | |
| 360.35 | |
| ZLBLYGIIADHDKG-UHFFFAOYSA-N | |
| naphthalene-1,5-disulfonic acid tetrahydrate |
| C10H16O10S2 | |
| MFCD00149265 | |
| 53249422 | |
| O.O.O.O.OS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(O)(=O)=O |
Specifications
| 211366-30-2 | |
| C10H16O10S2 | |
| 25 g | |
| ZLBLYGIIADHDKG-UHFFFAOYSA-N | |
| naphthalene-1,5-disulfonic acid tetrahydrate | |
| 53249422 | |
| ≥98.0% (HPLC,T) | |
| 1,5-Naphthalenedisulfonic Acid Tetrahydrate |
| White-Yellow | |
| MFCD00149265 | |
| 3261 | |
| O.O.O.O.OS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(O)(=O)=O | |
| 360.35 | |
| 288.29 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0467
RTECSNumber : QJ5900000
TSCA : No