accessibility menu, dialog, popup

UserName

SOOHOSOOHOH2OH2OH2OH2O

CAS RN 211366-30-2

IUPAC Name: naphthalene-1,5-disulfonic acid tetrahydrate
Synonyms: 1,5-naphthalenedisulfonic acid tetrahydrate
Molecular Weight (g/mol): 360.35
Molecular Formula: C10H16O10S2
InChi Key: ZLBLYGIIADHDKG-UHFFFAOYSA-N
SMILES: O.O.O.O.OS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results