missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Hexadiene 97.0+%, TCI America™
Supplier: TCI America H0084100ML
Specifications
| 1,5-Hexadiene | |
| Colorless | |
| C6H10 | |
| 100 mL | |
| 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl | |
| C=CCCC=C | |
| 82.146 | |
| 82.15 | |
| Liquid |
| 592-42-7 | |
| 59°C | |
| MFCD00008666 | |
| 2458 | |
| PYGSKMBEVAICCR-UHFFFAOYSA-N | |
| hexa-1,5-diene | |
| 11598 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 592-42-7 | |
| 82.146 | |
| PYGSKMBEVAICCR-UHFFFAOYSA-N | |
| 11598 | |
| C=CCCC=C |
| C6H10 | |
| MFCD00008666 | |
| 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl | |
| hexa-1,5-diene |
Safety and Handling
EINECSNumber : (2)-3694
RTECSNumber : MM1997800
TSCA : Yes