missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiene 97.0+%, TCI America™

Catalog No. p-7130325
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H008425ML 25 mL
H0084100ML 100 mL
H0084500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H008425ML Supplier TCI America Supplier No. H008425ML
Only null left

Chemical Identifiers

CAS 592-42-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008666
InChI Key PYGSKMBEVAICCR-UHFFFAOYSA-N
Synonym 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl
PubChem CID 11598
IUPAC Name hexa-1,5-diene
SMILES C=CCCC=C

Specifications

Color Colorless
Boiling Point 59°C
Quantity 25 mL
UN Number 2458
Formula Weight 82.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiene
EINECSNumber (2)-3694
RTECSNumber MM1997800
TSCA Yes