accessibility menu, dialog, popup

UserName

H2CCH2

CAS RN 592-42-7

IUPAC Name: hexa-1,5-diene
Synonyms: 1,5-hexadiene
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: PYGSKMBEVAICCR-UHFFFAOYSA-N
SMILES: C=CCCC=C
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)