missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diphenylcarbohydrazide, ACS, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D1063100GM
Specifications
| 140-22-7 | |
| 173° to 176°C | |
| Amber Glass Bottle | |
| 100 g | |
| O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1 | |
| 242.28 | |
| Crystalline Powder or Lumps |
| 1 | |
| 0.0005 | |
| C13H14N4O | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| 1,3-bis(phenylamino)urea | |
| ACS |
Chemical Identifiers
| 140-22-7 | |
| 242.28 | |
| 1,3-bis(phenylamino)urea |
| C13H14N4O | |
| KSPIHGBHKVISFI-UHFFFAOYSA-N | |
| O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label Safety Data Sheet (SDS) prior to use.