missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbohydrazide, ACS, Spectrum™ Chemical

Catalog No. p-7165465
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608061 25 g Amber Glass Bottle
18608062 100 g Amber Glass Bottle
18608063 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608061 Supplier Spectrum Chemical Mfg Cor Supplier No. D106325GM
Only null left

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.28
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
IUPAC Name 1,3-bis(phenylamino)urea
SMILES O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1

Specifications

CAS Max % 1
Melting Point 173° to 176°C
Ignition Residue 0.0005
Packaging Amber Glass Bottle
Quantity 25 g
Grade ACS
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label Safety Data Sheet (SDS) prior to use.