missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dinitronaphthalene, 97+%

Catalog No. AAA1386914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1386914 25 g
AAA1386922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1386914 Supplier Thermo Scientific Chemicals Supplier No. A1386914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 605-71-0 | C10H6N2O4 | 218.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 605-71-0
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD00003916
InChI Key ZUTCJXFCHHDFJS-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene
PubChem CID 11803
ChEBI CHEBI:50640
IUPAC Name 1,5-dinitronaphthalene
SMILES C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-]

Specifications

Melting Point 214°C to 218°C
Density 1.586
Odor Odorless
Quantity 25 g
Beilstein 527184
Formula Weight 218.17
Percent Purity ≥97%
Chemical Name or Material 1,5-Dinitronaphthalene
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 210-095-2
RTECSNumber QJ4551000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only