missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diiodo-2,4-dimethylbenzene 97.0+%, TCI America™

Catalog No. p-7128603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50051G 1 g
D50055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50051G Supplier TCI America Supplier No. D50051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4102-50-5 | C8H8I2 | 357.96 g/mol

Chemical Identifiers

CAS 4102-50-5
Molecular Formula C8H8I2
Molecular Weight (g/mol) 357.96
MDL Number MFCD29089354
InChI Key ZOOZGIAPEXFLEF-UHFFFAOYSA-N
Synonym 4,6-Diiodo-m-xylene
PubChem CID 12575960
IUPAC Name 1,5-diiodo-2,4-dimethylbenzene
SMILES CC1=CC(C)=C(I)C=C1I

Specifications

Melting Point 72°C
Quantity 1 g
Packaging Glass Bottle
Physical Form Crystal-Powder at 20°C
Formula Weight 357.96
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 1,5-Diiodo-2,4-dimethylbenzene