accessibility menu, dialog, popup

UserName

CH3IICH3

CAS RN 4102-50-5

IUPAC Name: 1,5-diiodo-2,4-dimethylbenzene
Synonyms: 1,5-diiodo-2,4-dimethylbenzene
Molecular Weight (g/mol): 357.96
Molecular Formula: C8H8I2
InChi Key: ZOOZGIAPEXFLEF-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(I)C=C1I
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results