missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloro-2,4-dinitrobenzene, 97%

CAS: 3698-83-7 | C6H2Cl2N2O4 | 236.992 g/mol

$300.10 - $300.10

Chemical Identifiers

CAS 3698-83-7
Molecular Formula C6H2Cl2N2O4
Molecular Weight (g/mol) 236.992
MDL Number MFCD00024186
InChI Key ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
PubChem CID 77286
IUPAC Name 1,5-dichloro-2,4-dinitrobenzene
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0665214
View Documents
Thermo Scientific Chemicals
L0665214
25 g
Each for $300.10
Only null left
 
Description

Description

1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloro-2,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3698-83-7
236.992
ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
77286
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]
C6H2Cl2N2O4
MFCD00024186
1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
1,5-dichloro-2,4-dinitrobenzene

Specifications

3698-83-7
C6H2Cl2N2O4
25 g
1685438
Insoluble in water.
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)[N+](=O)[O-]
236.992
237
1,5-Dichloro-2,4-dinitrobenzene
98°C to 103°C
MFCD00024186
UN2811
1,3-dichloro-4,6-dinitrobenzene, benzene, 1,5-dichloro-2,4-dinitro, 2,4-dichloro-1,5-dinitrobenzene, 4,6-dichloro-1,3-dinitrobenzene, unii-6enc7c8zr0, 6enc7c8zr0, 1,5-dichloro-2,4-dinitro-benzene, 1,3,-dichloro-4,6-dinitrobenzene, 1,5-dichlor-2,4-dinitrobenzol, 2,5-dinitrobenzene
ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
1,5-dichloro-2,4-dinitrobenzene
77286
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 223-027-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only