accessibility menu, dialog, popup

UserName

CAS RN 3698-83-7

IUPAC Name: 1,5-dichloro-2,4-dinitrobenzene
Synonyms: 1,5-dichloro-2,4-dinitrobenzene
Molecular Weight (g/mol): 236.99
Molecular Formula: C6H2Cl2N2O4
InChi Key: ZPXDNSYFDIHPOJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(Cl)C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)