missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dibromo-2,4-dimethylbenzene 97.0+%, TCI America™
Supplier: TCI America D500425G
Specifications
| 1,5-Dibromo-2,4-dimethylbenzene | |
| 615-87-2 | |
| 256°C | |
| Crystal-Powder at 20°C | |
| MFCD00154984 | |
| SOYPUUFPUFRXRI-UHFFFAOYSA-N | |
| 1,5-dibromo-2,4-dimethylbenzene | |
| 621990 | |
| ≥97.0% (GC) |
| 71°C | |
| White | |
| 25 g | |
| C8H8Br2 | |
| 4,6-Dibromo-m-xylene | |
| CC1=CC(C)=C(Br)C=C1Br | |
| 263.96 | |
| 263.96 |
Chemical Identifiers
| 615-87-2 | |
| 263.96 | |
| SOYPUUFPUFRXRI-UHFFFAOYSA-N | |
| 621990 | |
| CC1=CC(C)=C(Br)C=C1Br |
| C8H8Br2 | |
| MFCD00154984 | |
| 4,6-Dibromo-m-xylene | |
| 1,5-dibromo-2,4-dimethylbenzene |
Safety and Handling
TSCA : No