accessibility menu, dialog, popup

UserName

CH3BrBrCH3

CAS RN 615-87-2

IUPAC Name: 1,5-dibromo-2,4-dimethylbenzene
Synonyms: 1,5-dibromo-2,4-dimethylbenzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: SOYPUUFPUFRXRI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results