missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-2,4-dimethylbenzene 97.0+%, TCI America™

Catalog No. D50045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50045G 5 g
D500425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50045G Supplier TCI America Supplier No. D50045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-87-2 | C8H8Br2 | 263.96 g/mol

Chemical Identifiers

CAS 615-87-2
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00154984
InChI Key SOYPUUFPUFRXRI-UHFFFAOYSA-N
Synonym 4,6-Dibromo-m-xylene
PubChem CID 621990
IUPAC Name 1,5-dibromo-2,4-dimethylbenzene
SMILES CC1=CC(C)=C(Br)C=C1Br

Specifications

Melting Point 71°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 256°C
Formula Weight 263.96
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 1,5-Dibromo-2,4-dimethylbenzene