missing translation for 'onlineSavingsMsg'
Learn More

1,5,6,7-Tetrahydro-4H-indol-4-one 99.0+%, TCI America™

Supplier:  TCI America T143525G

 View more versions of this product

Catalog No. T143525G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,5,6,7-Tetrahydro-4H-indol-4-one
190°C
MFCD00075438
4-Oxo-4,5,6,7-tetrahydroindole
C1CC2=C(C=CN2)C(=O)C1
135.166
135.17
Crystalline Powder
13754-86-4
C8H9NO
25 g
KASJZXHXXNEULX-UHFFFAOYSA-N
1,5,6,7-tetrahydroindol-4-one
280229
≥99.0% (GC)

Chemical Identifiers

13754-86-4
135.166
KASJZXHXXNEULX-UHFFFAOYSA-N
280229
C1CC2=C(C=CN2)C(=O)C1
C8H9NO
MFCD00075438
4-Oxo-4,5,6,7-tetrahydroindole
1,5,6,7-tetrahydroindol-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions