accessibility menu, dialog, popup

UserName

CAS RN 13754-86-4

OHN

CAS RN 13754-86-4

IUPAC Name: 4,5,6,7-tetrahydro-1H-indol-4-one
Synonyms: 1,5,6,7-tetrahydroindol-4-one
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: KASJZXHXXNEULX-UHFFFAOYSA-N
SMILES: O=C1CCCC2=C1C=CN2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results