missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5,6,7-Tetrahydro-4H-indol-4-one 99.0+%, TCI America™
$34.66 - $102.03
Chemical Identifiers
| CAS | 13754-86-4 |
|---|---|
| Molecular Formula | C8H9NO |
| Molecular Weight (g/mol) | 135.166 |
| MDL Number | MFCD00075438 |
| InChI Key | KASJZXHXXNEULX-UHFFFAOYSA-N |
| Synonym | 4-Oxo-4,5,6,7-tetrahydroindole |
| PubChem CID | 280229 |
| IUPAC Name | 1,5,6,7-tetrahydroindol-4-one |
| SMILES | C1CC2=C(C=CN2)C(=O)C1 |
Chemical Identifiers
| 13754-86-4 | |
| 135.166 | |
| KASJZXHXXNEULX-UHFFFAOYSA-N | |
| 280229 | |
| C1CC2=C(C=CN2)C(=O)C1 |
| C8H9NO | |
| MFCD00075438 | |
| 4-Oxo-4,5,6,7-tetrahydroindole | |
| 1,5,6,7-tetrahydroindol-4-one |
Specifications
| 13754-86-4 | |
| C8H9NO | |
| 5 g | |
| KASJZXHXXNEULX-UHFFFAOYSA-N | |
| 1,5,6,7-tetrahydroindol-4-one | |
| 280229 | |
| ≥99.0% (GC) | |
| 1,5,6,7-Tetrahydro-4H-indol-4-one |
| 190°C | |
| MFCD00075438 | |
| 4-Oxo-4,5,6,7-tetrahydroindole | |
| C1CC2=C(C=CN2)C(=O)C1 | |
| 135.166 | |
| 135.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No